Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964223.1 | XP_027820024.1 | 12 - 13 | 2887 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |