Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015096389.2 | XP_014951875.2 | 1 - 2 | 541 | 88 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Ovis aries |