Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969306.1 | XP_027825107.1 | 7 - 8 | 762 | 64 | |
XM_027969307.1 | XP_027825108.1 | 6 - 7 | 717 | 64 | ||
XM_027969308.1 | XP_027825109.1 | 7 - 8 | 708 | 64 | ||
XM_027969309.1 | XP_027825110.1 | 7 - 8 | 729 | 64 | ||
XM_027969310.1 | XP_027825111.1 | 6 - 7 | 521 | 64 | ||
XM_027969312.1 | XP_027825113.1 | 5 - 6 | 496 | 64 | ||
XM_027969313.1 | XP_027825114.1 | 6 - 7 | 671 | 64 | ||
XM_027969314.1 | XP_027825115.1 | 5 - 6 | 476 | 64 | ||
XM_027969315.1 | XP_027825116.1 | 6 - 7 | 508 | 64 | ||
XM_027969316.1 | XP_027825117.1 | 5 - 6 | 496 | 64 | ||
XM_027969317.1 | XP_027825118.1 | 5 - 6 | 547 | 64 | ||
XM_027969318.1 | XP_027825119.1 | 6 - 7 | 666 | 64 | ||
XM_027969319.1 | XP_027825120.1 | 5 - 6 | 485 | 64 | ||
XM_027969320.1 | XP_027825121.1 | 7 - 8 | 566 | 64 | ||
XM_027969321.1 | XP_027825122.1 | 6 - 7 | 496 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |