Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104497.2 | XP_014959983.2 | 13 - 14 | 1907 | 78 | |
XM_027962238.1 | XP_027818039.1 | 14 - 15 | 1994 | 78 | ||
XM_027962239.1 | XP_027818040.1 | 14 - 15 | 1991 | 78 | ||
XM_027962240.1 | XP_027818041.1 | 14 - 15 | 1994 | 78 | ||
XM_027962241.1 | XP_027818042.1 | 14 - 15 | 1994 | 78 | ||
XM_027962242.1 | XP_027818043.1 | 14 - 15 | 1994 | 78 | ||
XM_027962243.1 | XP_027818044.1 | 14 - 15 | 1991 | 78 |