Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAB21L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973497.1 | XP_027829298.1 | 6 - 7 | 840 | 60 | |
XM_027973498.1 | XP_027829299.1 | 6 - 7 | 856 | 60 | ||
XM_027973499.1 | XP_027829300.1 | 3 - 4 | 603 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |