Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965844.1 | XP_027821645.1 | 4 - 5 | 754 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |