Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015097698.2 | XP_014953184.1 | 2 - 3 | 733 | 106 | |
XM_015097700.2 | XP_014953186.1 | 2 - 3 | 603 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |