Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956338.1 | XP_027812139.1 | 4 - 5 | 948 | 72 | |
XM_027956339.1 | XP_027812140.1 | 4 - 5 | 945 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |