Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964000.1 | XP_027819801.1 | 4 - 5 | 1468 | 71 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Ovis aries |