Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPZA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006807.4 | XP_004006856.1 | 1 | 462 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |