Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018442.4 | XP_004018491.1 | 2 - 3 | 1040 | 80 | |
XM_012100028.3 | XP_011955418.1 | 2 - 3 | 237 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |