Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967148.1 | XP_027822949.1 | 2 - 3 | 758 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |