Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006692.4 | XP_004006741.2 | 1 - 2 | 630 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |