Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCF24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011719.4 | XP_004011768.3 | 1 - 2 | 387 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |