Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974180.1 | XP_027829981.1 | 11 - 12 | 2319 | 76 | |
XM_027974181.1 | XP_027829982.1 | 11 - 12 | 2367 | 76 | ||
XM_027974182.1 | XP_027829983.1 | 10 - 11 | 2489 | 76 | ||
XM_027974183.1 | XP_027829984.1 | 11 - 12 | 2271 | 76 | ||
XM_027974185.1 | XP_027829986.1 | 12 - 13 | 2178 | 76 | ||
XM_027974186.1 | XP_027829987.1 | 12 - 13 | 1990 | 76 |