Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012190423.3 | XP_012045813.1 | 2 - 3 | 1310 | 73 | |
XM_027978293.1 | XP_027834094.1 | 1 - 2 | 2559 | 73 | ||
XM_027978294.1 | XP_027834095.1 | 2 - 3 | 1234 | 73 |