Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963423.1 | XP_027819224.1 | 7 - 8 | 1585 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |