Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012099781.3 | XP_011955171.1 | 3 - 4 | 313 | 115 | |
XM_012099782.3 | XP_011955172.1 | 3 - 4 | 343 | 115 | ||
XM_012099784.2 | XP_011955174.1 | 3 - 4 | 313 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |