Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016914.3 | XP_004016963.1 | 3 - 4 | 356 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |