Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFHR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976096.1 | XP_027831897.1 | 9 - 10 | 1474 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |