Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004023371.4 | XP_004023420.3 | 2 - 3 | 202 | 76 | |
XM_027977523.1 | XP_027833324.1 | 1 - 2 | 491 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |