Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004023371.4 | XP_004023420.3 | 5 - 6 | 745 | 82 | |
XM_027977523.1 | XP_027833324.1 | 4 - 5 | 1034 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |