Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015103786.2 | XP_014959272.2 | 3 - 4 | 1383 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |