Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005776.4 | XP_004005825.1 | 3 - 4 | 1192 | 107 | |
XM_012166878.3 | XP_012022268.1 | 4 - 5 | 1270 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |