Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012104999.3 | XP_011960389.1 | 3 - 4 | 542 | 79 | |
XM_012105001.3 | XP_011960391.1 | 3 - 4 | 409 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |