Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976506.1 | XP_027832307.1 | 2 - 3 | 198 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |