Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASRGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019598.4 | XP_004019647.2 | 6 - 7 | 870 | 57 | |
GenBank mRNA | EU366480.1 | - | 1 - 2 | 104 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |