Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSEN34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978854.1 | XP_027834655.1 | 3 - 4 | 609 | 107 | |
XM_027978855.1 | XP_027834656.1 | 3 - 4 | 595 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |