Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKX2-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963900.1 | XP_027819701.1 | 1 - 2 | 279 | 190 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |