Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MINPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020010.4 | XP_004020059.3 | 3 - 4 | 933 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |