Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXRA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963681.1 | XP_027819482.1 | 4 - 5 | 937 | 78 | |
XM_027963682.1 | XP_027819483.1 | 4 - 5 | 797 | 78 |