Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGF29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012104338.3 | XP_011959728.1 | 9 - 10 | 931 | 78 | |
XM_012104339.3 | XP_011959729.1 | 10 - 11 | 915 | 78 |