Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092305.2 | XP_014947791.2 | 6 - 7 | 1567 | 104 | |
XM_027976425.1 | XP_027832226.1 | 6 - 7 | 1567 | 104 | ||
XM_027976429.1 | XP_027832230.1 | 6 - 7 | 1564 | 104 | ||
XM_027976433.1 | XP_027832234.1 | 7 - 8 | 1432 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |