Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010414.3 | XP_004010463.1 | 1 - 2 | 132 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |