Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011413.4 | XP_004011462.2 | 4 - 5 | 811 | 69 | |
XM_012183113.3 | XP_012038503.2 | 3 - 4 | 731 | 69 | ||
XM_012183116.3 | XP_012038506.1 | 2 - 3 | 217 | 69 | ||
XM_012183117.3 | XP_012038507.1 | 2 - 3 | 186 | 69 | ||
XM_027972562.1 | XP_027828363.1 | 4 - 5 | 816 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |