Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012180163.3 | XP_012035553.2 | 1 - 2 | 264 | 99 | |
XM_012180164.3 | XP_012035554.2 | 1 - 2 | 264 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |