Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016511.4 | XP_004016560.2 | 5 - 6 | 685 | 87 | |
XM_027979825.1 | XP_027835626.1 | 5 - 6 | 657 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |