Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012098549.3 | XP_011953939.1 | 1 - 2 | 79 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |