Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966306.1 | XP_027822107.1 | 1 - 2 | 281 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |