Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101101851 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015091798.2 | XP_014947284.2 | 2 | 391 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |