Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014588.4 | XP_004014637.1 | 3 - 4 | 637 | 113 | |
XM_012189228.3 | XP_012044618.1 | 3 - 4 | 370 | 113 | ||
XM_012189229.3 | XP_012044619.1 | 3 - 4 | 264 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |