Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019435.4 | XP_004019484.1 | 1 - 2 | 626 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |