Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015093523.2 | XP_014949009.2 | 2 - 3 | 602 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |