Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP27X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022600.3 | XP_004022649.2 | 2 | 2186 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |