Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SACS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973664.1 | XP_027829465.1 | 3 - 4 | 343 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |