Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF3C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015094120.2 | XP_014949606.2 | 4 - 5 | 2403 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |