Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC114110138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960006.1 | XP_027815807.1 | 2 | 461 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |