Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195836.1 | NP_001182765.1 | 9 - 10 | 1389 | 70 | |
XM_008266001.2 | XP_008264223.1 | 9 - 10 | 1646 | 70 | ||
XM_008266002.2 | XP_008264224.1 | 8 - 9 | 1271 | 70 | ||
XM_017346624.1 | XP_017202113.1 | 8 - 9 | 1270 | 70 | ||
XM_017346625.1 | XP_017202114.1 | 8 - 9 | 1232 | 70 | ||
GenBank mRNA | AF149419.1 | - | 1 - 2 | 157 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |