Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136522.1 | NP_001129994.1 | 25 - 26 | 3641 | 65 | |
GenBank mRNA | AF199511.1 | - | 11 - 12 | 1079 | 65 | |
X15539.1 | - | 3641 | 65 | |||
X55763.1 | - | 3979 | 65 | |||
X58695.1 | - | 372 | 65 | |||
X58696.1 | - | 372 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus Inventoried Probe spans exons |