Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008267633.2 | XP_008265855.1 | 42 - 43 | 6625 | 67 | |
XM_008267635.2 | XP_008265857.1 | 42 - 43 | 6610 | 67 | ||
XM_008267636.2 | XP_008265858.1 | 41 - 42 | 6058 | 67 | ||
XM_008267637.2 | XP_008265859.1 | 39 - 40 | 6052 | 67 | ||
XM_008267638.2 | XP_008265860.1 | 39 - 40 | 6037 | 67 | ||
XM_017347439.1 | XP_017202928.1 | 41 - 42 | 6514 | 67 | ||
XM_017347440.1 | XP_017202929.1 | 34 - 35 | 5348 | 67 | ||
GenBank mRNA | AJ286819.1 | - | 4 - 5 | 277 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |